Biphenyl-2,2'-dicarboxylic anhydride ≥98.0%

Supplier: TCI America

Synonyms: Diphenic anhydride

D0865-5G D0865-25G
TCD0865-5GEA 222.78 CAD
TCD0865-5G TCD0865-025G
Biphenyl-2,2'-dicarboxylic anhydride ≥98.0%
Biphenyl-2,2'-dicarboxylic anhydrid

CAS Number: 6050-13-1
MDL Number: MFCD00004993
Molecular Formula: C14H8O3
Molecular Weight: 224.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 230


Other Product Names / Synonyms: 2,2'-Bibenzoic Anhydride, 2,2'-Biphenyldicarboxylic Anhydride

Formula: C₁₄H₈O₃
MW: 224.22 g/mol
Melting Pt: 230 °C
MDL Number: MFCD00004993
CAS Number: 6050-13-1
EINECS: 227-950-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e20767652003d511e560f1d085fdc5f825a9768&vid=4e20767652003d511e560f1d085fdc5f825a9768&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR