Biphenyl-2,2'-dicarboxylic acid 98,0

Supplier: TCI America

Synonyms: [1,1'-Biphenyl]-2,2'-dicarboxylic acid, Diphenic acid, 2,2'-Biphenyldicarboxylic acid

D0864-25G D0864-100G
TCD0864-025GEA 462.94 CAD
TCD0864-025G TCD0864-100G
Biphenyl-2,2'-dicarboxylic acid 98,0
Biphenyl-2,2'-dicarboxylic acid

CAS Number: 482-05-3
MDL Number: MFCD00002464
Molecular Formula: C14H10O4
Molecular Weight: 242.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 233


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents, Monomers, Biphenyls (for High-Performance Polymer Research)
Other Product Names / Synonyms: 2,2'-Bibenzoic Acid, 2,2'-Dicarboxybiphenyl, Diphenic Acid

Formula: C₁₄H₁₀O₄
MW: 242.23 g/mol
Melting Pt: 233 °C
MDL Number: MFCD00002464
CAS Number: 482-05-3
EINECS: 207-576-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99157a515a09b6bbc0b274f21bcc1e790f586cf1&vid=99157a515a09b6bbc0b274f21bcc1e790f586cf1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR