Nitro blue tetrazolium chloride (NBT) ≥98.0% (by HPLC, titration analysis) for biochemical research

Supplier: TCI America
Warning

Synonyms: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride

D0844-1G D0844-100MG
TCD0844-001GEA 244.3 CAD
TCD0844-001G TCD0844-100MG
Nitro blue tetrazolium chloride (NBT) ≥98.0% (by HPLC, titration analysis) for biochemical research
Nitro blue tetrazolium chloride (NBT)

[for Biochemical Research]
CAS Number: 298-83-9
MDL Number: MFCD00012159
Molecular Formula: C40H30Cl2N10O6
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 223
Storage Temperature: <0°C


Other Product Names / Synonyms: 2,2'-Di-p-nitrophenyl-5,5'-diphenyl-(3,3'-dimethoxy)-4,4'-bisphenyleneditetrazolium Chloride, NBT

Formula: C₄₀H₃₀Cl₂N₁₀O₆
Melting Pt: 223 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012159
CAS Number: 298-83-9
EINECS: 206-067-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e42e2c7ffb64be9f45938a1a20a23635f505685&vid=74bb7e7999e5c709cb9252ce6cb01a235e0c3e7f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR