3,5-Dinitrobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

D0824-500G D0824-25G
TCD0824-500GEA 137.46 CAD
TCD0824-500G TCD0824-025G
3,5-Dinitrobenzoic acid ≥98.0% (by GC, titration analysis)
3,5-Dinitrobenzoic acid

CAS Number: 99-34-3
MDL Number: MFCD00007253
Molecular Formula: C7H4N2O6
Molecular Weight: 212.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 207

Formula: C₇H₄N₂O₆
Melting Pt: 207 °C
Density: 1.683 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007253
CAS Number: 99-34-3
EINECS: 202-751-1
UN: 1325
ADR: 4.1,III
Merck Index: 13,03304
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a736220b1516b22f50ae2faeba3a27dfbca6a4b&vid=d077c1438ff6dc245eaedbd5d08e2e0aa18d33d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR