3,4-Dinitrobenzoic acid ≥98.0%

Supplier: TCI America
Warning

D0823-5G D0823-25G
TCD0823-005GEA 75.57 CAD
TCD0823-005G TCD0823-025G
3,4-Dinitrobenzoic acid ≥98.0%
3,4-Dinitrobenzoic acid

CAS Number: 528-45-0
MDL Number: MFCD00007255
Molecular Formula: C7H4N2O6
Molecular Weight: 212.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 165

Formula: C₇H₄N₂O₆
MW: 212.12 g/mol
Melting Pt: 165 °C
Density: 1.674 g/cm³ (20 °C)
MDL Number: MFCD00007255
CAS Number: 528-45-0
EINECS: 208-433-9
Merck Index: 13,03303
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db8ff7071533ac99651df6fd1322faf4eb6ccf7e&vid=46dd35ac6eb5e8042f74ace909e6ef6dbba4647f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR