2,3-Dimethylquinoxaline ≥98.0%

Supplier: TCI America
Warning

D0795-100G D0795-25G
TCD0795-100GEA 406.06 CAD
TCD0795-100G TCD0795-025G
2,3-Dimethylquinoxaline ≥98.0%
2,3-Dimethylquinoxaline

CAS Number: 2379-55-7
MDL Number: MFCD00006728
Molecular Formula: C10H10N2
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 107

Formula: C₁₀H₁₀N₂
Boiling Pt: 130 °C
Melting Pt: 107 °C
Density: 1.226 g/cm³ (20 °C)
MDL Number: MFCD00006728
CAS Number: 2379-55-7
EINECS: 219-162-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e94951f780f94f0d73f6fa40dd779b25055145f&vid=4e94951f780f94f0d73f6fa40dd779b25055145f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR