2,6-Dimethyl-4-pyrone ≥99.0%

Supplier: TCI America

Synonyms: 2,6-Dimethyl-4H-pyran-4-one, 2,6-Dimethyl-γ-pyrone

D0794-25G D0794-5G
TCD0794-025GEA 322.32 CAD
TCD0794-025G TCD0794-5G
2,6-Dimethyl-4-pyrone ≥99.0%
2,6-Dimethyl-4-pyrone

CAS Number: 1004-36-0
MDL Number: MFCD00006579
Molecular Formula: C7H8O2
Molecular Weight: 124.14
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 134


Other Product Names / Synonyms: 2,6-Dimethyl-4H-pyran-4-one

Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 134 °C
Density: 1.17 g/cm³ (25 °C)
MDL Number: MFCD00006579
CAS Number: 1004-36-0
EINECS: 213-719-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7dd480c23fbf69375145a65a6472814b6dcfff51&vid=7dd480c23fbf69375145a65a6472814b6dcfff51&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR