2,2-Dimethyl-1,3-propanediol ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Neopentyl Glycol, 2,2-Dimethylpropane-1,3-diol
D0791-25G
D0791-500G
TCD0791-25GEA
28.28
CAD
TCD0791-25G
TCD0791-500G
2,2-Dimethyl-1,3-propanediol ≥98.0%
2,2-Dimethyl-1,3-propanediol
CAS Number: 126-30-7
MDL Number: MFCD00004685
Molecular Formula: C5H12O2
Molecular Weight: 104.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 210
Melting point (°C): 128
Flash Point (°C): 107
Other Product Names / Synonyms: 2,2-Bis(hydroxymethyl)propane, 2,2-Dihydroxy-2,2-dimethylpropane, Neopentyl Glycol
Formula:
C₅H₁₂O₂ MW: 104.15 g/mol Boiling Pt: 208 Melting Pt: 128 °C Density: 1.07 g/cm³ (20 °C) Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004685 CAS Number: 126-30-7 EINECS: 204-781-0 Merck Index: 12,06547 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...