2-Methyl-1-phenyl-2-propanol ≥98.0%

Supplier: TCI America
Warning

D0783-500ML D0783-25ML D0783-100ML
TCD0783-500MLEA 142.2 CAD
TCD0783-500ML TCD0783-025ML TCD0783-100ML
2-Methyl-1-phenyl-2-propanol ≥98.0%

CAS Number: 100-86-7
MDL Number: MFCD00004465
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 94
Flash Point (°C): 92
Freezing point (°C): 23


Other Product Names / Synonyms: Benzyldimethylcarbinol, alpha,alpha-Dimethylphenethyl Alcohol

Formula: C₆H₅CH₂C(CH₃)(OH)CH₃
Boiling Pt: 94 °C
Melting Pt: 23…25 °C
Density: 0.974 g/cm³ (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00004465
CAS Number: 100-86-7
EINECS: 202-896-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac777db219c13ec81e86506d935e238c9a78e573&vid=ac777db219c13ec81e86506d935e238c9a78e573&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR