N,N-Dimethyl-4-nitroaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D0760-25G D0760-5G
TCD0760-025GEA 391.84 CAD
TCD0760-025G TCD0760-005G
N,N-Dimethyl-4-nitroaniline ≥98.0% (by GC, titration analysis)
N,N-Dimethyl-4-nitroaniline

CAS Number: 100-23-2
MDL Number: MFCD00014712
Molecular Formula: C8H10N2O2
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 165

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Boiling Pt: 288 °C (1013 hPa)
Melting Pt: 165 °C
Density: 1.179 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014712
CAS Number: 100-23-2
EINECS: 202-832-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f26a5e2ec7cb582b90cdae966193a93e1332f8e&vid=aef66f17b092728d4367ebd845c0a944f464cb81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR