N,N-Dimethyl-1-naphthylamine ≥99.0%

Supplier: TCI America
Warning

Synonyms: Dimethyl(1-naphthyl)amine, N,N-Dimethylnaphthalen-1-amine

D0755-500G D0755-25G
TCD0755-500GEA 2637.02 CAD
TCD0755-500G TCD0755-25G
N,N-Dimethyl-1-naphthylamine ≥99.0%
N,N-Dimethyl-1-naphthylamine

CAS Number: 86-56-6
MDL Number: MFCD00003919
Molecular Formula: C12H13N
Molecular Weight: 171.24
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 274
Flash Point (°C): 140
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: 1-Dimethylaminonaphthalene

Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Boiling Pt: 275
Density: 1.04 (20 °C)
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00003919
CAS Number: 86-56-6
EINECS: 201-682-4
UN: 3082
ADR: 9 III
Merck Index: 12,03301
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b791419ef9f06465233256e7dbf718bd99609f68&vid=b791419ef9f06465233256e7dbf718bd99609f68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR