5,5-Dimethylhydantoin ≥98.0%

Supplier: TCI America

Synonyms: 5,5-Dimethylimidazolidine-2,4-dione

D0739-500G D0739-25G
TCD0739-500GEA 75.84 CAD
TCD0739-500G TCD0739-025G
5,5-Dimethylhydantoin ≥98.0%
5,5-Dimethylhydantoin

CAS Number: 77-71-4
MDL Number: MFCD00005266
Molecular Formula: C5H8N2O2
Molecular Weight: 128.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 175
Flash Point (°C): 193

Formula: C₅H₈N₂O₂
Melting Pt: 175 °C
Flash Pt: 193 °C
MDL Number: MFCD00005266
CAS Number: 77-71-4
EINECS: 201-051-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c8e0516d66499f64d8c9f749f5071536fecf1c2&vid=6c8e0516d66499f64d8c9f749f5071536fecf1c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR