2,5-Dimethyl-3-hexyne-2,5-diol ≥99.0%
Supplier: TCI America
D0738-25G
D0738-500G
TCD0738-025GEA
36.96
CAD
TCD0738-025G
TCD0738-500G
2,5-Dimethyl-3-hexyne-2,5-diol ≥99.0%
2,5-Dimethyl-3-hexyne-2,5-diol
CAS Number: 142-30-3
MDL Number: MFCD00004468
Molecular Formula: C8H14O2
Molecular Weight: 142.20
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 206
Melting point (°C): 95
Flash Point (°C): 110
Other Product Names / Synonyms: 2,5-Dihydroxy-2,5-dimethyl-3-hexyne
Formula:
C₈H₁₄O₂ Boiling Pt: 206 °C Melting Pt: 95 °C Density: 0.9469 g/cm³ (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00004468 CAS Number: 142-30-3 EINECS: 205-533-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...