Dimethyl glyoxime ≥98.0% (by gravimetric analysis)

Supplier: TCI America
Danger

Synonyms: Diacetyl glyoxime, Diacetyl dioxime, 2,3-Butanedione dioxime

D0729-25G D0729-500G
TCD0729-025GEA 45.01 CAD
TCD0729-025G TCD0729-500G
Dimethyl glyoxime ≥98.0% (by gravimetric analysis)
Dimethyl glyoxime

CAS Number: 95-45-4
MDL Number: MFCD00002117
Molecular Formula: C4H8N2O2
Molecular Weight: 116.12
Purity/Analysis Method: >98.0% (W)
Form: Crystal
Melting point (°C): 240


Other Product Names / Synonyms: 2,3-Butanedione Dioxime, Diacetyl Dioxime

Formula: CH₃C(=NOH)C(=NOH)CH₃
MW: 116.12 g/mol
Boiling Pt: 53 °C (8 mmHg)
Melting Pt: 240 °C
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002117
CAS Number: 95-45-4
EINECS: 202-420-1
Merck Index: 13,03272
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d0fc7693a5413fb54cd6bd135fc940ef715022e&vid=5d0fc7693a5413fb54cd6bd135fc940ef715022e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (W)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR