Cetyldimethylethylammonium bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Ethylhexadecyldimethylammonium Bromide, Hexadecylethyldimethylammonium bromide, Ethyl(1-hexadecyl)dimethylammonium bromide, Ethylhexadecyldimethylammonium bromide, Dimethylethylhexadecylammonium bromide

D0721-25G D0721-500G
TCD0721-025GEA 52.14 CAD
TCD0721-025G TCD0721-500G
Cetyldimethylethylammonium bromide ≥98.0% (by titrimetric analysis)
Cetyldimethylethylammonium bromide

CAS Number: 124-03-8
MDL Number: MFCD00011774
Molecular Formula: C20H44BrN
Molecular Weight: 378.48
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Cetylethyldimethylammonium Bromide

Formula: C₂₀H₄₄BrN
MW: 378.4741 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011774
CAS Number: 124-03-8
EINECS: 204-672-8
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4301330f948a42a0de576b934e1747da8952f65&vid=b4301330f948a42a0de576b934e1747da8952f65&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR