(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol ≥98.0%
Supplier: TCI America
D0710-500G
D0710-25G
D0710-100G
TCD0710-500GEA
336.54
CAD
TCD0710-500G
TCD0710-025G
TCD0710-100G
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol ≥98.0%
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
CAS Number: 100-79-8
MDL Number: MFCD00063238
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 80
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: 2,2-Dimethyl-4-hydroxymethyl-1,3-dioxolane, 1,2-Isopropylideneglycerol, Solketal
Formula:
C₆H₁₂O₃ Boiling Pt: 80 °C Density: 1.07 (20 °C) Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063238 CAS Number: 100-79-8 EINECS: 202-888-7 Merck Index: 13,05233 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...