Dimedone ≥99.0% (by titrimetric analysis)

Supplier: TCI America

D0701-25G D0701-500G D0701-100G
TCD0701-025GEA 75.52 CAD
TCD0701-025G TCD0701-500G TCD0701-100G
Dimedone ≥99.0% (by titrimetric analysis)
Dimedone

CAS Number: 126-81-8
MDL Number: MFCD00001588
Molecular Formula: C8H12O2
Molecular Weight: 140.18
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 149


Other Product Names / Synonyms: 5,5-Dimethyl-1,3-cyclohexanedione, 5,5-Dimethyldihydroresorcinol, Methone

Formula: C₈H₁₂O₂
Boiling Pt: 234 °C (1013 hPa)
Melting Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00001588
CAS Number: 126-81-8
EINECS: 204-804-4
Merck Index: 13,03268
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db51dff55c6cf1b485bf4a374bd24edb042c3c56&vid=db51dff55c6cf1b485bf4a374bd24edb042c3c56&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR