4,4'-Dimethylbenzophenone ≥99.0%

Supplier: TCI America

D0685-25G D0685-5G
TCD0685-025GEA 197.5 CAD
TCD0685-025G TCD0685-005G
4,4'-Dimethylbenzophenone ≥99.0%
4,4'-Dimethylbenzophenone

CAS Number: 611-97-2
MDL Number: MFCD00017214
Molecular Formula: C15H14O
Molecular Weight: 210.28
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 151
Melting point (°C): 95


Other Product Names / Synonyms: Di-p-tolyl Ketone

Formula: C₁₅H₁₄O
Boiling Pt: 151 °C
Melting Pt: 95 °C
Density: 1.18 g/cm³ (20 °C)
MDL Number: MFCD00017214
CAS Number: 611-97-2
EINECS: 210-287-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b433b564fce1d8e7c767263d3cc7a6ee3ced79c9&vid=b433b564fce1d8e7c767263d3cc7a6ee3ced79c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR