3,5-Dimethylbenzoic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

D0684-25G D0684-250G
TCD0684-025GEA 107.44 CAD
TCD0684-025G TCD0684-250G
3,5-Dimethylbenzoic acid ≥99.0% (by titrimetric analysis)
3,5-Dimethylbenzoic acid

CAS Number: 499-06-9
MDL Number: MFCD00002525
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 170


Other Product Names / Synonyms: Mesitylenic Acid, m-Xylene-5-carboxylic Acid

Formula: C₉H₁₀O₂
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 170 °C
Density: 1.211 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002525
CAS Number: 499-06-9
EINECS: 207-876-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d793f80c24849e07cf74f3ff7cd6ce35cbea4b9&vid=ce108e06f4644bf5e99c2208a9a6b51c51c0c930&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR