Methyl-2-(dimethylamino)benzoate ≥97.0%

Supplier: TCI America

Synonyms: 2-(Dimethylamino)benzoic acid methyl ester

D0675-25G D0675-10G
TCD0675-025GEA 83.74 CAD
TCD0675-025G TCD0675-010G
Methyl-2-(dimethylamino)benzoate ≥97.0%
Methyl 2-(dimethylamino)benzoate

CAS Number: 10072-05-6
MDL Number: MFCD00025858
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 131
Flash Point (°C): 122
Specific Gravity (20/20): 1.10


Other Product Names / Synonyms: N,N-Dimethylanthranilic Acid Methyl Ester, Methyl 2-(Dimethylamino)benzoate

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Boiling Pt: 131 °C
Density: 1.10 (20 °C)
Flash Pt: 122 °C
MDL Number: MFCD00025858
CAS Number: 10072-05-6
EINECS: 233-202-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87a8eceeb0351b1eea51206dff65b91db749929b&vid=87a8eceeb0351b1eea51206dff65b91db749929b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR