3-(Dimethylamino)phenol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N,N-Dimethyl-3-hydroxyaniline, 3-Dimethylaminophenol

D0657-25G D0657-100G
TCD0657-025GEA 185.71 CAD
TCD0657-025G TCD0657-100G
3-(Dimethylamino)phenol ≥97.0% (by GC, titration analysis)
3-(Dimethylamino)phenol

CAS Number: 99-07-0
MDL Number: MFCD00002264
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 268
Melting point (°C): 86
Flash Point (°C): 80


Other Product Names / Synonyms: N,N-Dimethyl-3-aminophenol

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 268 °C
Melting Pt: 86 °C
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00002264
CAS Number: 99-07-0
EINECS: 202-727-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7c54ba1db3bf1c57018aaac7f9a8ca095a8f8ee&vid=d7c54ba1db3bf1c57018aaac7f9a8ca095a8f8ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR