4-(Dimethylamino)cinnamaldehyde ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D0648-5G D0648-25G
TCD0648-005GEA 141.92 CAD
TCD0648-005G TCD0648-025G
4-(Dimethylamino)cinnamaldehyde ≥98.0% (by titrimetric analysis)
4-(Dimethylamino)cinnamaldehyde

CAS Number: 6203-18-5
MDL Number: MFCD00007002
Molecular Formula: C11H13NO
Molecular Weight: 175.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 140


Product Categories: Bioscience, Phenylpropanoids, Analytical Science, Absolute Configuration Determination (Exciton Chirality CD Method)
Other Product Names / Synonyms: 3-[4-(Dimethylamino)phenyl]acrylaldehyde, 3-[4-(Dimethylamino)phenyl]-2-propenal

Formula: C₁₁H₁₃NO
Melting Pt: 140 °C
MDL Number: MFCD00007002
CAS Number: 6203-18-5
EINECS: 228-267-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7172a0b4cf7c70f300c237de2a2be7487e2c9db&vid=a7172a0b4cf7c70f300c237de2a2be7487e2c9db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR