5-(4-(Dimethylamino)benzylidene)rhodanine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D0647-25G
D0647-1G
TCD0647-025GEA
772.62
CAD
TCD0647-025G
TCD0647-001G
5-(4-(Dimethylamino)benzylidene)rhodanine ≥98.0% (by HPLC, titration analysis)
5-(4-(Dimethylamino)benzylidene)rhodanine
CAS Number: 536-17-4
MDL Number: MFCD00064857
Molecular Formula: C12H12N2OS2
Molecular Weight: 264.36
Purity/Analysis Method:
Other Product Names / Synonyms: 4-Dimethylaminobenzalrhodanine, 5-(4-Dimethylaminobenzylidene)-2-thioxo-4-thiazolidinone
Formula:
C₁₂H₁₂N₂OS₂ Melting Pt: 288 °C |
MDL Number:
MFCD00064857 CAS Number: 536-17-4 EINECS: 208-625-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...