3-(Dimethylamino)benzoic acid ≥98.0%

Supplier: TCI America

D0646-25G D0646-500G
TCD0646-025GEA 52.14 CAD
TCD0646-025G TCD0646-500G
3-(Dimethylamino)benzoic acid ≥98.0%
3-(Dimethylamino)benzoic acid

CAS Number: 99-64-9
MDL Number: MFCD00002497
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 151

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 298
Melting Pt: 151 °C
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002497
CAS Number: 99-64-9
EINECS: 202-775-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfb6080d6a59ad9416ff52a1bb5cdd5c1051eab0&vid=22838e72d37528e793c0ba27eb7ebe658ffd1213&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR