2,3-Dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D0630-25G D0630-500G
TCD0630-025GEA 69.52 CAD
TCD0630-025G TCD0630-500G
2,3-Dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
2,3-Dimethoxybenzoic acid

CAS Number: 1521-38-6
MDL Number: MFCD00002432
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 124

Formula: C₉H₁₀O₄
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 124 °C
Density: 1.387 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002432
CAS Number: 1521-38-6
EINECS: 216-188-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d24e46ee239fd144f3ca3e528b2bdd45cb48f70&vid=1fdfbd8a01ce5363f433bdd7bd858449faccc59f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR