1,4-Dimethoxybenzene ≥99.0%

Supplier: TCI America

D0629-25G D0629-500G
TCD0629-025GEA 45.82 CAD
TCD0629-025G TCD0629-500G
1,4-Dimethoxybenzene ≥99.0%
1,4-Dimethoxybenzene

CAS Number: 150-78-7
MDL Number: MFCD00008401
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 212
Melting point (°C): 56
Flash Point (°C): 88


Other Product Names / Synonyms: Hydroquinone Dimethyl Ether

Formula: C₈H₁₀O₂
Boiling Pt: 212 °C
Melting Pt: 56 °C
Density: 0.874 g/cm³ (24.9 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00008401
CAS Number: 150-78-7
EINECS: 205-771-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb39aea87d4d9e5d69a9d69b8bf6d2763be0de95&vid=b51f792e191b1fbe2c734fcf56068c68c590b0f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR