1,3-Dimethoxybenzene ≥99.0%
Supplier: TCI America
D0628-25G
D0628-500G
D0628-100G
TCD0628-025GEA
40.2
CAD
TCD0628-025G
TCD0628-500G
TCD0628-100G
1,3-Dimethoxybenzene ≥99.0%
1,3-Dimethoxybenzene
CAS Number: 151-10-0
MDL Number: MFCD00008384
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 217
Flash Point (°C): 87
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: Resorcinol Dimethyl Ether
Formula:
C₈H₁₀O₂ Boiling Pt: 217 °C Melting Pt: –52 °C Density: 1.07 (20 °C) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008384 CAS Number: 151-10-0 EINECS: 205-783-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...