2,3-Dimethoxybenzaldehyde ≥98.0%
Supplier: TCI America
D0625-500G
D0625-25G
TCD0625-500GEA
677.82
CAD
TCD0625-500G
TCD0625-025G
2,3-Dimethoxybenzaldehyde ≥98.0%
2,3-Dimethoxybenzaldehyde
CAS Number: 86-51-1
MDL Number: MFCD00003309
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 256
Melting point (°C): 53
Flash Point (°C): 110
Formula:
C₉H₁₀O₃ Boiling Pt: 256 °C Melting Pt: 53 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003309 CAS Number: 86-51-1 EINECS: 201-677-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...