1,4-Diiodobenzene ≥98.0%

Supplier: TCI America

D0608-25G D0608-5G
TCD0608-25GEA 113.76 CAD
TCD0608-25G TCD0608-005G
1,4-Diiodobenzene ≥98.0%
1,4-Diiodobenzene

CAS Number: 624-38-4
MDL Number: MFCD00001054
Molecular Formula: C6H4I2
Molecular Weight: 329.91
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 129

Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 129 °C
Density: 2.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001054
CAS Number: 624-38-4
EINECS: 210-842-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc831b0926b2316e3910874662adb03f98fa0e19&vid=f4c5190f8bc3dfa72bb5b2aa45df8dc6efe2d717&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR