1,3-Diiodobenzene ≥99.0%
Supplier: TCI America
|
Warning
|
D0607-5G
D0607-25G
TCD0607-005GEA
42.66
CAD
TCD0607-005G
TCD0607-25G
1,3-Diiodobenzene ≥99.0%
1,3-Diiodobenzene
CAS Number: 626-00-6
MDL Number: MFCD00041731
Molecular Formula: C6H4I2
Molecular Weight: 329.91
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 284
Melting point (°C): 35
Flash Point (°C): 110
Formula:
C₆H₄I₂ MW: 329.91 g/mol Boiling Pt: 284 °C Melting Pt: 35 °C Density: 2.76 g/cm³ (0 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00041731 CAS Number: 626-00-6 EINECS: 210-921-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...