1,2-Diiodobenzene ≥98.0% stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with Stabilised

D0606-25G D0606-5G
TCD0606-25GEA 799.95 CAD
TCD0606-25G TCD0606-005G
1,2-Diiodobenzene ≥98.0% stabilized
1,2-Diiodobenzene

(stabilized with Copper chip)
CAS Number: 615-42-9
MDL Number: MFCD00001038
Molecular Formula: C6H4I2
Molecular Weight: 329.91
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 152
Flash Point (°C): 110
Freezing point (°C): 23
Storage Temperature: 0-10°C

Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 152 °C
Density: 2.76 g/cm³ (0 °C)
Flash Pt: 110 °C
MDL Number: MFCD00001038
CAS Number: 615-42-9
EINECS: 210-425-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60f555c076b2d2ba390417713d3855e93b2ef44e&vid=914579d64d3fe130f38f7f160e8dc571cfffff02&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR