4-(Hydroxymethyl)benzyl alcohol ≥99.0%

Supplier: TCI America

Synonyms: p-Phenylenedimethanol, 1,4-Benzenedimethanol, 1,4-Bis(hydroxymethyl)-benzene, p-Xylene-α,α'-diol

D0605-500G D0605-25G D0605-100G
TCD0605-500GEA 457.39 CAD
TCD0605-500G TCD0605-025G TCD0605-100G
4-(Hydroxymethyl)benzyl alcohol ≥99.0%
4-(Hydroxymethyl)benzyl alcohol

CAS Number: 589-29-7
MDL Number: MFCD00004665
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 143
Melting point (°C): 118
Flash Point (°C): 186


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: p-Xylylene Glycol, alpha,alpha'-Dihydroxy-p-xylene, p-Xylene-alpha,alpha'-diol

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 143 °C
Melting Pt: 118 °C
Flash Pt: 186 °C
MDL Number: MFCD00004665
CAS Number: 589-29-7
EINECS: 209-641-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64efb5370ee46d1cca111192a39cdba1bcd238fa&vid=64efb5370ee46d1cca111192a39cdba1bcd238fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR