2,7-Dihydroxynaphthalene ≥99.0%

Supplier: TCI America
Warning

D0594-25G D0594-500G D0594-100G
TCD0594-025GEA 76.79 CAD
TCD0594-025G TCD0594-500G TCD0594-100G
2,7-Dihydroxynaphthalene ≥99.0%
2,7-Dihydroxynaphthalene

CAS Number: 582-17-2
MDL Number: MFCD00004085
Molecular Formula: C10H8O2
Molecular Weight: 160.17
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 186


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,7-Naphthalenediol

Formula: C₁₀H₈O₂
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 186 °C
Storage Temperature: Ambient
MDL Number: MFCD00004085
CAS Number: 582-17-2
EINECS: 209-478-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8374b746307ca163aee6b979346836218201e4e7&vid=8374b746307ca163aee6b979346836218201e4e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR