2,3-Dihydroxynaphthalene ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D0593-250G D0593-25G
TCD0593-250GEA 594.08 CAD
TCD0593-250G TCD0593-025G
2,3-Dihydroxynaphthalene ≥98.0% (by GC, titration analysis)
2,3-Dihydroxynaphthalene

CAS Number: 92-44-4
MDL Number: MFCD00004073
Molecular Formula: C10H8O2
Molecular Weight: 160.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 163


Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes, Monomers
Other Product Names / Synonyms: 2,3-Naphthalenediol

Formula: C₁₀H₈O₂
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 163 °C
Density: 1.12 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004073
CAS Number: 92-44-4
EINECS: 202-156-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=134ae5164be2a1eb4aef2b4b13e914f600acad54&vid=134ae5164be2a1eb4aef2b4b13e914f600acad54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR