1,7-Dihydroxynaphthalene ≥98.0%

Supplier: TCI America
Warning

Synonyms: Naphthalene-1,7-diol

D0592-500G D0592-25G
TCD0592-500GEA 1717.46 CAD
TCD0592-500G TCD0592-025G
1,7-Dihydroxynaphthalene ≥98.0%
1,7-Dihydroxynaphthalene

CAS Number: 575-38-2
MDL Number: MFCD00035720
Molecular Formula: C10H8O2
Molecular Weight: 160.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 175
Flash Point (°C): 252
Lambda max.: 335 nm (Cyclohexane)


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,7-Naphthalenediol

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 175 °C
Flash Pt: 252 °C
Storage Temperature: Ambient
MDL Number: MFCD00035720
CAS Number: 575-38-2
EINECS: 209-383-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=068d76e1c726d7956179a765780bc9122a6ec3ea&vid=068d76e1c726d7956179a765780bc9122a6ec3ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR