1,5-Dihydroxynaphthalene ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1,5-Naphthalenediol, Naphthalene-1,5-diol
D0590-25G
D0590-500G
D0590-100G
TCD0590-025GEA
53.01
CAD
TCD0590-025G
TCD0590-500G
TCD0590-100G
1,5-Dihydroxynaphthalene ≥98.0%
1,5-Dihydroxynaphthalene
CAS Number: 83-56-7
MDL Number: MFCD00003980
Molecular Formula: C10H8O2
Molecular Weight: 160.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,5-Naphthalenediol
Formula:
C₁₀H₈O₂ MW: 160.17 g/mol Boiling Pt: 375 Melting Pt: 180 Density: 1.41 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003980 CAS Number: 83-56-7 EINECS: 201-487-4 UN: 3077 ADR: 9 III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...