1,3-Dihydroxynaphthalene ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D0588-1G D0588-5G
TCD0588-001GEA 105.88 CAD
TCD0588-001G TCD0588-5G
1,3-Dihydroxynaphthalene ≥98.0% (by GC, titration analysis)
1,3-Dihydroxynaphthalene

CAS Number: 132-86-5
MDL Number: MFCD00003965
Molecular Formula: C10H8O2
Molecular Weight: 160.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 125


Other Product Names / Synonyms: 1,3-Naphthalenediol, Naphthoresorcinol

Formula: C₁₀H₈O₂
Boiling Pt: 386 °C (1013 hPa)
Melting Pt: 125 °C
Density: 1.12 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003965
CAS Number: 132-86-5
EINECS: 205-079-7
Merck Index: 12,06483
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85a838ccf838b096bd242fe09d0dc92cfb6250a6&vid=85a838ccf838b096bd242fe09d0dc92cfb6250a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR