2,4-Diamino-6-methyl-1,3,5-triazine ≥98.0%

Supplier: TCI America

Synonyms: 2,4-Diamino-6-methyl-1,3,5-triazine 6-Methyl-1,3,5-triazine-2,4-diamine, 2,4-Diamino-6-methyl-s-triazine, 6-Methyl-1,3,5-triazine-2,4-diamine, Acetoguanamine, 6-Methyl-1,3,5-triazine-2,4-diyldiamine

D0583-25G D0583-500G
TCD0583-025GEA 33.18 CAD
TCD0583-025G TCD0583-500G
2,4-Diamino-6-methyl-1,3,5-triazine ≥98.0%
2,4-Diamino-6-methyl-1,3,5-triazine

CAS Number: 542-02-9
MDL Number: MFCD00023192
Molecular Formula: C4H7N5
Molecular Weight: 125.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acetoguanamine

Formula: C₄H₇N₅
MW: 125.13 g/mol
MDL Number: MFCD00023192
CAS Number: 542-02-9
EINECS: 208-796-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df4cbd36684371919c52130f27c6b161c1a94eb1&vid=df4cbd36684371919c52130f27c6b161c1a94eb1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR