Ethyl-3,4-dihydroxybenzoate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3,4-Dihydroxybenzoic acid ethyl ester, Protocatechuic acid ethyl ester, Ethyl 3,4-dihydroxybenzoate

D0571-5G D0571-25G
TCD0571-5GEA 45.82 CAD
TCD0571-5G TCD0571-025G
Ethyl-3,4-dihydroxybenzoate ≥98.0% (by GC, titration analysis)
Ethyl 3,4-dihydroxybenzoate

CAS Number: 3943-89-3
MDL Number: MFCD00002199
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 133


Other Product Names / Synonyms: 3,4-Dihydroxybenzoic Acid Ethyl Ester, Ethyl Protocatechuate, Protocatechuic Acid Ethyl Ester

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Melting Pt: 133 °C
MDL Number: MFCD00002199
CAS Number: 3943-89-3
EINECS: 223-529-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9612e016a4e8bd69f6cf7b3ca4ced2c370b6b6ae&vid=9612e016a4e8bd69f6cf7b3ca4ced2c370b6b6ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR