2,5-Dihydroxybenzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D0569-25G D0569-500G
TCD0569-025GEA 99.38 CAD
TCD0569-025G TCD0569-500G
2,5-Dihydroxybenzoic acid ≥98.0% (by HPLC, titration analysis)
2,5-Dihydroxybenzoic acid

CAS Number: 490-79-9
MDL Number: MFCD00002460
Molecular Formula: C7H6O4
Molecular Weight: 154.12
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 206


Other Product Names / Synonyms: Gentisic Acid

Formula: C₇H₆O₄
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 206 °C
Density: 1.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002460
CAS Number: 490-79-9
EINECS: 207-718-5
Merck Index: 13,04409
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3be5e2c1fccc422476662fda5f1cf3feec5d3139&vid=579ffa6688289b130ef0722b01c0131d8034af22&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR