2,5-Dihydroxybenzaldehyde ≥98.0%

Supplier: TCI America
Warning

D0565-1G D0565-5G
TCD0565-001GEA 44.24 CAD
TCD0565-001G 76218-960
2,5-Dihydroxybenzaldehyde ≥98.0%
2,5-Dihydroxybenzaldehyde

CAS Number: 1194-98-5
MDL Number: MFCD00003333
Molecular Formula: C7H6O3
Molecular Weight: 138.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 102


Other Product Names / Synonyms: Gentisinaldehyde

Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 60…65 °C (12 mmHg)
Melting Pt: 102 °C
Density: 0.859 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003333
CAS Number: 1194-98-5
EINECS: 214-789-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=855a2dbaad9683046e88381a51b4ee6ad3baefef&vid=d5a41990cb475d61a1e133879b01d9cf10aa9917&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR