3-Sulfolene ≥98.0%

Supplier: TCI America

Synonyms: 3-sulfolene, 3-sulpholene, Butadiene sulfone, Butadiene sulphone

D0557-25G D0557-500G
TCD0557-025GEA 39.22 CAD
TCD0557-025G TCD0557-500G
3-Sulfolene ≥98.0%
3-Sulpholene

CAS Number: 77-79-2
MDL Number: MFCD00005481
Molecular Formula: C4H6O2S
Molecular Weight: 118.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 65


Product Categories: Synthetic Organic Chemistry, Sulfones
Other Product Names / Synonyms: 2,5-Dihydrothiophene 1,1-Dioxide

Formula: C₄H₆O₂S
Melting Pt: 65 °C
MDL Number: MFCD00005481
CAS Number: 77-79-2
EINECS: 201-059-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e45360f3b1487cd38747206a26545f197c6851be&vid=e45360f3b1487cd38747206a26545f197c6851be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR