Diglycolic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D0545-500G
D0545-25G
TCD0545-500GEA
562.48
CAD
TCD0545-500G
TCD0545-025G
Diglycolic acid ≥98.0% (by GC, titration analysis)
Diglycolic acid
CAS Number: 110-99-6
MDL Number: MFCD00004309
Molecular Formula: C4H6O5
Molecular Weight: 134.09
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 144
Other Product Names / Synonyms: Oxydiacetic Acid
Formula:
C₄H₆O₅ Melting Pt: 144 °C |
MDL Number:
MFCD00004309 CAS Number: 110-99-6 EINECS: 203-823-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...