Bis(4-fluorophenyl) sulfone ≥99.0%
Supplier: TCI America
Synonyms:
1-Fluor-4-((4-fluorphenyl)sulfonyl)benzol, 4-Fluorophenyl sulphone, 4,4'-Sulfonylbis(fluorobenzene), 4-Fluorophenyl sulfone, 4,4'-Difluoro diphenyl sulphone, 4,4'-Difluoro diphenyl sulfone, 1-Fluor-4-((4-fluorphenyl)sulphonyl)benzol, 4,4'-sulphonylbis(fluorobenzene)
CAS Number: 383-29-9
MDL Number: MFCD00000350
Molecular Formula: C12H8F2O2S
Molecular Weight: 254.25
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 99
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Diphenyl Sulfones (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Difluorodiphenyl Sulfone
Formula:
C₁₂H₈F₂O₂S MW: 254.26 g/mol Melting Pt: 99 °C |
MDL Number:
MFCD00000350 CAS Number: 383-29-9 EINECS: 206-847-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...