1,4-Diethoxybenzene ≥98.0%
Supplier: TCI America
D0535-25G
D0535-500G
TCD0535-025GEA
56.25
CAD
TCD0535-025G
TCD0535-500G
1,4-Diethoxybenzene ≥98.0%
1,4-Diethoxybenzene
CAS Number: 122-95-2
MDL Number: MFCD00015146
Molecular Formula: C10H14O2
Molecular Weight: 166.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 112
Melting point (°C): 70
Other Product Names / Synonyms: Hydroquinone Diethyl Ether
Formula:
C₁₀H₁₄O₂ Boiling Pt: 112 °C Melting Pt: 70 °C |
MDL Number:
MFCD00015146 CAS Number: 122-95-2 EINECS: 204-585-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...