1,3-Diethylurea ≥98.0% (by GC, total nitrogen)
Supplier: TCI America
D0534-25G
D0534-500G
TCD0534-025GEA
237
CAD
TCD0534-025G
TCD0534-500G
1,3-Diethylurea ≥98.0% (by GC, total nitrogen)
1,3-Diethylurea
CAS Number: 623-76-7
MDL Number: MFCD00009028
Molecular Formula: C5H12N2O
Molecular Weight: 116.16
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 111
Formula:
C₅H₁₂N₂O Melting Pt: 111 °C |
MDL Number:
MFCD00009028 CAS Number: 623-76-7 EINECS: 210-811-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...