1,3-Diethyl-2-thiourea ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N,N'-Diethylthiourea

D0530-25G D0530-500G
TCD0530-025GEA 26.86 CAD
TCD0530-025G TCD0530-500G
1,3-Diethyl-2-thiourea ≥97.0% (by titrimetric analysis)
1,3-Diethyl-2-thiourea

CAS Number: 105-55-5
MDL Number: MFCD00004925
Molecular Formula: C5H12N2S
Molecular Weight: 132.23
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 79

Formula: C₅H₁₂N₂S
MW: 132.23 g/mol
Boiling Pt: 169
Melting Pt: 79 °C
Density: 1.118 g/cm³ (20 °C)
Flash Pt: 186
MDL Number: MFCD00004925
CAS Number: 105-55-5
EINECS: 203-308-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f351375c7731d3a6f56e3fb5571a975e8048e02b&vid=f351375c7731d3a6f56e3fb5571a975e8048e02b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR