N,N-Diethyl-p-phenylenediammonium sulfate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Amino-N,N-diethylaniline sulfate, N,N-Diethyl-p-phenylenediamine sulphate, N,N-Diethyl-1,4-phenylenediammonium sulphate, N1,N1-Diethylbenzene-1,4-diamine sulfate, 4-Amino-N,N-diethylaniline sulphate, N,N-Diethyl-1,4-phenylenediammonium sulfate, N,N-Diethyl-p-phenylenediamine sulfate, N,N-Diethylbenzene-1,4-diammonium sulfate, N1,N1-Diethylbenzene-1,4-diamine sulphate, N,N-Diethylbenzene-1,4-diammonium sulphate

D0520-25G D0520-500G D0520-100G
TCD0520-025GEA 53.72 CAD
TCD0520-025G TCD0520-500G TCD0520-100G
N,N-Diethyl-p-phenylenediammonium sulfate ≥98.0% (by HPLC, titration analysis)
N,N-Diethyl-p-phenylenediammonium sulphate

CAS Number: 6283-63-2
MDL Number: MFCD00012993
Molecular Formula: C10H16N2
Molecular Weight: 262.32
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 185


Other Product Names / Synonyms: 4-Amino-N,N-diethylaniline Sulfate

Formula: C₁₀H₁₈N₂O₄S
MW: 262.33 g/mol
Boiling Pt: 274…275 °C (1013 hPa)
Melting Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00012993
CAS Number: 6283-63-2
EINECS: 228-500-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6aa14ed1ab3cc23e3e032de82475dc59854a7e99&vid=6aa14ed1ab3cc23e3e032de82475dc59854a7e99&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR