Veratraldehyde ≥98.0%

Supplier: TCI America
Warning

D0507-25G D0507-500G D0507-100G
TCD0507-025GEA 48.81 CAD
TCD0507-025G TCD0507-500G TCD0507-100G
Veratraldehyde ≥98.0%
Veratraldehyde

CAS Number: 120-14-9
MDL Number: MFCD00003363
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 281
Melting point (°C): 43


Other Product Names / Synonyms: Veratraldehyde

Formula: C₉H₁₀O₃
Boiling Pt: 281 °C
Melting Pt: 43 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003363
CAS Number: 120-14-9
EINECS: 204-373-2
Merck Index: 12,10084
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94e7b5794c81ff0d70bf6056cb3ceefd901bc40c&vid=288e461417391faf1a9b591f620b8d1d2c655790&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR