Diethylene glycol monohexyl ether ≥98.0%

Supplier: TCI America

Synonyms: 2-(2-(Hexyloxy)ethoxy)ethanol, Diethylene glycol mono-n-hexyl ether

D0501-25ML D0501-500ML
TCD0501-025MLEA 26.86 CAD
TCD0501-025ML TCD0501-500ML
Diethylene glycol monohexyl ether ≥98.0%
Diethylene glycol monohexyl ether

CAS Number: 112-59-4
MDL Number: MFCD00010703
Molecular Formula: C10H22O3
Molecular Weight: 190.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 258
Flash Point (°C): 141
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: Hexyl Carbitol, 2-(2-Hexyloxyethoxy)ethanol

Formula: C₁₀H₂₂O₃
MW: 190.28 g/mol
Boiling Pt: 258 °C
Density: 0.94 (20 °C)
Flash Pt: 141 °C
MDL Number: MFCD00010703
CAS Number: 112-59-4
EINECS: 203-988-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6449fb609dfe562c1c51ac42bbb1e68aa809f055&vid=6449fb609dfe562c1c51ac42bbb1e68aa809f055&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR